missing translation for 'onlineSavingsMsg'
Learn More

Ansell Profile 3000 Nitrile Examination Gloves

Product Code. 12970825
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Combine comfort and chemical resistance with Nitritex™ Profile 3000 nitrile exam gloves. Ambidextrous, latex-free glove offers chemical and puncture resistance.

  • Nitrile is resistant to a wide range of chemicals
  • Excellent puncture resistance
  • Beaded cuff provides easier donning and stability
  • Textured surface for increased grip
  • Powder-free and latex-free to eliminate Type I latex allergies
  • Wrist-length gloves measure 240mm/9.5 in.
  • Easy-access 100-glove dispenser box
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 9001:2008, 13485:2003 and ISO 2859
Classification Class I Medical Device
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Examination Glove
Size 3X-Large
Sterility Non-sterile
For Use With (Application) Premium examinations
Glove Cuff Style Beaded/Rolled
Inspected for Medical Use Yes
Model Profile 3000
Packaging Type Boxed
Quantity 100/Pk.
Series Profile 3000
Length (Metric) 24 cm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'