missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WTW™ ProfiLine™ Portable Conductivity Meters
Obtain optimum measurements in natural waters and wastewater with this reliable conductivity portable meter.
1225.00 €
Specifications
Accuracy (Conductivity) | ±0.5% Full-scale ±1 LSD |
---|---|
For Use With (Application) | For professional field measurements requiring documentation and data transfer |
Cell Constant | Fixed: 0.475cm–1/1cm–1, Calibratable: 0.450 to 0.500cm–1/0.585 to 715cm–1/0.800 to 1.200cm–1 |
Calibration Points | 1, 2, 3-point WTW Technical and NIST Buffer |
Range (Conductivity) | 0 μS to 1000 mS |
Product Code | Brand | Calibration Points | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Calibration Points | Includes | Price | Quantity & Availability | ||||
10414885
|
WTW™
2CA101 |
1, 2, 3-point WTW Technical and NIST Buffer | Meter with carry case including 4-electrode conductivity cell TetraCon™ 325, standard solution 1413μS/cm at 25°C, stand, beaker, short instruction manual, CD-ROM and batteries |
1225.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Robust and waterproof meter for portable conductivity measurement
- Reliable conductivity meter with automatic nIF temperature compensation according to DIN EN 27888 for routine measurements in natural waters and wastewater
- Features automatic temperature compensation nlF
- Includes meter with carry case including 4-electrode conductivity cell TetraCon™ 325, standard solution 1413μS/cm at 25°C, stand, beaker, short instruction manual, CD-ROM and batteries
- For professional field measurements requiring documentation and data transfer
Specifications
±0.5% Full-scale ±1 LSD | |
Fixed: 0.475cm–1/1cm–1, Calibratable: 0.450 to 0.500cm–1/0.585 to 715cm–1/0.800 to 1.200cm–1 | |
0 μS to 1000 mS | |
Automatic | |
0.1 ppt | |
0.1 | |
LCD | |
IP67 | |
3 Years | |
4 x 1.5V AA oder 4 x 1.2V NiMH-rechargeable battery | |
Portable Conductivity Meter | |
7.9 cm | |
NiMH |
For professional field measurements requiring documentation and data transfer | |
1, 2, 3-point WTW Technical and NIST Buffer | |
0.001, 0.01, 0.1 mS | |
0 to 70 ppt | |
±0.2% Full-scale | |
±0.1°C | |
CE | |
0.6 kg | |
Cond 3110 | |
-5°C to +105°C | |
17.7 cm | |
5.7 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title