missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ ProFill HPLC Syringe Filters, Regenerated Cellulose Membrane

HPLC performance tested

Brand:  Fisherbrand™ 30 16 2089

261.39 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.


 View more versions of this product

Product Code. 10270225

  • 286.00 € / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

  • ProFill
  • HPLC Syringe Filter
  • Regenerated Cellulose (RC)
  • Low extractable membranes and housing
  • Polypropylene housing
  • Enhanced Luer Lock inlet which prevents leakage
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Brown
Regenerated Cellulose
4.91 cm2
PE-Bag, additionally packed in a PP-Box
6.2bar
30 mm
HPLC Syringe Filter
0.45 μm
<0.137 mL
No
100
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers