missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Progranulin (GRN) Mouse ELISA Kit

Sandwich ELISA Kit

614.00 € - 4815.00 €

Specifications

Accession Number P28798
Assay Sensitivity 20 pg/mL
Assay Solid-phase sandwich ELISA
Conjugate Biotin
Product Type ELISA Kit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18507824
View Documents Promotion Details
Invitrogen™
EMGRNX10
10 x 96 Tests
4815.00 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
15445273
View Documents Promotion Details
Invitrogen™
EMGRN
96 Tests
614.00 €
1 set
Estimated Shipment: 12-11-2024
to see stock.
Add to basket
 
Description

Description

The Mouse Progranulin (GRN) ELISA quantitates Ms Progranulin in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms Progranulin. Principle of the method The Mouse Progranulin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Progranulin is the precursor protein of Granulin also called as Proepithelin and PC cell-derived growth factor. Cleavage of the signal peptide produces mature Granulin. Both Granulin and smaller cleavage product of Granulin regulates cell growth. Progranulin stimulates proliferation of many epithelial cells, promotes their anchorage-independent growth and, when overexpressed, confers epithelial tumorigenicity. Progranulin/epithelin gene is multifunctional, with important constitutive roles in epithelial homeostasis, reproductive, immunological, and neuronal function. Progranulin constitutively expressed in a number of epithelia, particularly in the skin, GI tract, and reproductive system. Progranulin is expressed in immune cells in vivo and in specific neurons in the brain.
Specifications

Specifications

P28798
Solid-phase sandwich ELISA
ELISA Kit
96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
14824
<12%
HRP
Development
RP23-226P4.4, Pgrn, epithelin
1 hr. 20 min.
20 pg/mL
Biotin
Transcription Factors
Mouse
Colorimetric Microplate Reader
Grn
<10%
RUO
2°C to 8°C
Mouse
4 hr. 45 min.
Videos
SDS
Documents

Documents

Product Certifications