missing translation for 'onlineSavingsMsg'
Learn More

Proguanil hydrochloride, 97%, Thermo Scientific Chemicals

65.12 € - 195.00 €

Chemical Identifiers

CAS 637-32-1
Molecular Formula C11H16ClN5·HCl
Molecular Weight (g/mol) 290.19
InChI Key SARMGXPVOFNNNG-UHFFFAOYSA-N
Synonym proguanil hydrochloride, diguanyl, paludrine, proguanil hcl, chlorguanide hydrochloride, chloroquanil, bigumalum, chloroguanide hydrochloride, tirian hydrochloride, palusil hydrochloride
PubChem CID 9570076
IUPAC Name (1E)-1-[amino-(4-chloroanilino)methylidene]-2-propan-2-ylguanidine;hydrochloride
SMILES CC(C)N=C(N)N=C(N)NC1=CC=C(C=C1)Cl.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15668775
View Documents
Thermo Scientific Acros
461430250
25 mg
65.12 €
25mg
Please to purchase this item. Need a web account? Register with us today!
15658735
View Documents
Thermo Scientific Acros
461431000
100 mg
195.00 €
100mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

637-32-1
290.19
proguanil hydrochloride, diguanyl, paludrine, proguanil hcl, chlorguanide hydrochloride, chloroquanil, bigumalum, chloroguanide hydrochloride, tirian hydrochloride, palusil hydrochloride
(1E)-1-[amino-(4-chloroanilino)methylidene]-2-propan-2-ylguanidine;hydrochloride
C11H16ClN5·HCl
SARMGXPVOFNNNG-UHFFFAOYSA-N
9570076
CC(C)N=C(N)N=C(N)NC1=CC=C(C=C1)Cl.Cl

Specifications

245.0°C to 250.0°C
Authentic
C11H16ClN5·HCl
SARMGXPVOFNNNG-UHFFFAOYSA-N
(1E)-1-[amino-(4-chloroanilino)methylidene]-2-propan-2-ylguanidine;hydrochloride
9570076
97%
637-32-1
96% min. (HPLC)
proguanil hydrochloride, diguanyl, paludrine, proguanil hcl, chlorguanide hydrochloride, chloroquanil, bigumalum, chloroguanide hydrochloride, tirian hydrochloride, palusil hydrochloride
CC(C)N=C(N)N=C(N)NC1=CC=C(C=C1)Cl.Cl
290.19
290.19
Proguanil hydrochloride
Videos
Safety and Handling

Safety and Handling

Toxic if swallowed

Acute toxicity (category 3)

missing translation for 'einecsNumber' : 211-283-7

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.