Learn More
Sartorius Proline Plus Mechanical Pipette Multipacks
Multipacks combine a set of Proline Plus single-channel mechanical pipettes with matching tip trays and useful accessories. Sartorius Proline Pus Mechanical Pipette Multipacks provides best alternative to replace your used mechanical pipettes.
Brand: Sartorius LH-728674
999.28 EUR valid until 2024-12-31
Use promo code "21904" to get your promotional price.
Includes: 5 x Single-channel pipets (0.5 to 10μL, 10 to 100μL, 20 to 200μL, 100 to 1000μL, 500 to 5000μL), Tip Racks to 96 tips (0.1 to 10μL, 0.5 to 200μL, 10 to 1000μL), Tip Rack to 50 tips (5000μL), 1 x Linear Stand and 5 x Pipet Holder
Description
- Low pipetting forces
- Ergonomic finger hook
- Easy volume setting with click stop mechanism
- Quick and easy-to-read volume display minimises risk of volume setting errors
- Thermal insulation of internal components improves accuracy
- Easy to use “do-it-yourself” calibration
- Easy to clean with only three parts to disassemble
- Fully autoclavable
- High chemical and UV-resistance
- High-quality standard Optifit tips for various applications
- Optiload tip loading mechanism allows tip loading with an equal constant force onto every channel and enables perfect tip sealing onto every individual tip cone
Services
Two years
Specifications
Optifit tips | |
High chemical and UV resistant | |
1 | |
Non-sterile | |
0.5 to 10, 10 to 100, 20 to 200, 100 to 1000, 500 to 5000 μL | |
5 x Single-channel pipets (0.5 to 10μL, 10 to 100μL, 20 to 200μL, 100 to 1000μL, 500 to 5000μL), Tip Racks to 96 tips (0.1 to 10μL, 0.5 to 200μL, 10 to 1000μL), Tip Rack to 50 tips (5000μL), 1 x Linear Stand and 5 x Pipet Holder | |
UV resistant |
Autoclavable | |
Proline Plus | |
Proline Plus | |
Mechanical Pipette Multipacks | |
Safe-Cone Filter | |
10/Pk. | |
2 Years |