missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ ProLong™ Diamond Antifade Mountant

When sample thickness and imaging depth is not of concern, ProLong Diamond Antifade Mountant provides superior photobleach protection for Alexa Fluor dyes and fluorescent proteins allowing long term imaging experiments without loss of signal.

Brand:  Molecular Probes™ P36961

 View more versions of this product

Product Code. 15205739

  • 348.00 € / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

ProLong Diamond Antifade Mountant is a glycerol-based liquid mountant applied directly to fluorescently labeled cells or tissue samples. ProLong Diamond mountant is designed to provide unparalleled antifade protection across the entire visual and IR spectrums. You can use ProLong Diamond mountant with almost any fluorescent dye or fluorescent protein (e.g., GFP, RFP, mCherry) to achieve the brightest signal and lowest background, for bright, high-definition fluorescent images.

ProLong Diamond Antifade Mountant comes mixed and ready-to-use in a 2 mL dropper bottle or 10 mL bottle. Just apply a drop to the sample, add a coverslip, cure, and image. ProLong Diamond mountant does not discolor or shrink when cured, enabling high-quality fluorescent images months after mounting your sample. ProLong Diamond mountant will cure to an RI of 1.47.

Get photobleaching protection across the entire visible and IR spectrums
ProLong Diamond Antifade Mountant’s ability to provide brighter signal, against low background, helps ensure that you get high-resolution images. ProLong Diamond provides superior protection against photobleaching and will not quench your fluorophores. It can be used with an array of fluorescent dyes, including Alexa Fluor dyes, FITC, TRTC, Texas Red, Cy3, and Cy5 or fluorescent proteins. For immediate viewing of the sample without curing, SlowFade Diamond Antifade Mountant is optimal.

Archiving fluorescently labeled samples with excellent RI
ProLong Diamond mountant is the ideal option for archiving samples with fluorescent dyes or fluorescent proteins. Please note that ProLong Glass mountants are recommended for samples > 10 microns and all immersion oil imaging applications. It is also ideal for preserving signal strength during prolonged imaging sessions, such as repeated imaging under confocal laser scanning illumination, or with super resolution microscopy. Additionally, the curing property of ProLong Diamond means you do not need to use traditional acrylic resins (i.e., fingernail polish), which can damage microscope objectives if it comes into contact with them. The refractive index determines how much the path of light is bent, or refracted, when entering a material. In microscopy, refractive index differences between specimen and mounting medium should be minimized and can be reduced by using mountants with a higher RI. Optimizing the RI of your experiment will increase the resolution to provide better quality images. Depending on your sample type, thickness, imaging depth and/or staining reagents (e.g. fluorescent proteins or dyes) and whether you need a curing or non-curing mountant, choosing the right mounting media will impact image resolution and should be strongly considered for optimal experimental design.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 x 2 mL
Contains 5 dropper bottles contaning 2 mL reagent each.

• Store at 2-8°C.
• Protect from light.
• Stable for at least 6 months after receiving.
Mounting Solution, Antifade Solution
ProLong™ Diamond Antifade Mountant, 5 x 2 mL
Antifade Mountant
ProLong™
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.