missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ ProLong™ Live Antifade Reagent, for live cell imaging

Prolong Live Antifade Reagent helps prevent the loss of fluorescent signal (photobleaching) during live cell imaging.

Brand:  Molecular Probes™ P36974

200.64 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15421204

  • 264.00 € / Pack of 5
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

ProLong Live Antifade Reagent helps prevent the loss of fluorescent signal due to photobleaching during live cell imaging through use of enzymes from the plasma membrane of E. coli. These enzymes metabolize the elements that can cause photobleaching. When dyes are illuminated, they can degrade which reduces the sample’s fluorescent intensity and leads to the creation of free radical singlet oxygen which degrades neighboring dye molecules. ProLong Live Antifade Reagent metabolizes this element, increasing the signal intensity and duration while keeping the background signal low. ProLong Live reagent has little-to-no effect on cellular viability, proliferation, or other cellular functions in experiments up to 48 hours. ProLong Live reagent provides photobleach protection in live cell experiments for fluorophores such as GFP, RFP, Hoechst 33342, MitoTracker™, LysoTracker™, and CellTracker™dyes. ProLong Live Antifade Reagent can be added to any cell culture media or buffer that is suitable for fluorescent imaging.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 x 1 mL
Provided in 5 vials, with 1 mL each. Enough for 5000 assays (0.1 mL imaging solution/assay), or 1000 assays (0.5 mL imaging solution/assay). When stored at ≤-20°C product is stable for at least 6 months, with up to 4 freeze-thaw cycles. When stored at 2–8°C product should be used within 30 days (a precipitant may form at this temperature).
ProLong™
1 mL
Live Antifade Reagent
Approved for shipment at Room Temperature or on Wet Ice
Cell Imaging Reagent
ProLong™ Live Antifade Reagent for live cell imaging, 5 x 1 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.