Learn More
Heidolph™ Promax 1020
Temperature-controlled model with a load capacity of 5 kg.
Brand:  Heidolph™ 543-22332-06
Product Code. 17323030
  • 1995.00 € / Each
Estimated Shipment: 22-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The 1000 series reciprocating shaker can be easily integrated into the modular incubator system. This makes it possible to successfully run temperature controlled applications. The Promax 1020 can also be used to purify proteins at 4°C. With a platform size of 290 x 258 mm, there is enough space for the vessels used.
  • With a stroke of 32 mm and a speed of 30 to 250 rpm, a proper separation of the samples is guaranteed. The integrated timer function enables unattended operation so that you can concentrate on other tasks without worries. After the set time has elapsed, the acoustic signal signals the end of the shaking process.
  • Supplemented with the appropriate holder for separatory funnels, a maximum of four pieces between 50 and 100 mL can be securely attached to the Promax 1020.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
230 V
UK Plug
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Heidolph™ Promax 1020 > UK Plug