missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Promethazine hydrochloride

Product Code. 15740849
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-33-3
Molecular Formula C17H20N2S·HCl
Molecular Weight (g/mol) 320.88
InChI Key XXPDBLUZJRXNNZ-UHFFFAOYSA-N
Synonym promethazine hydrochloride, phenergan, promethazine hcl, fenergan, n,n-dimethyl-1-10h-phenothiazin-10-yl propan-2-amine hydrochloride, diprasine, farganesse, fellozine, pipolfen, pipolphen
PubChem CID 6014
ChEBI CHEBI:8462
IUPAC Name N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
SMILES CC(CN1C2=CC=CC=C2SC3=CC=CC=C31)N(C)C.Cl

Specifications

Melting Point 227.0°C to 230.0°C
Quantity 5 g
Solubility Information (10% in water) Clear colorless to light yellow
Formula Weight 320.88
Chemical Name or Material Promethazine hydrochloride
compliance-icons compliance-icons
Product Identifier
  • Promethazine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Promethazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'