missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Prong Clamp

With moving jaw pivoted for accurate alignment.

Brand:  Fisherbrand™ 2023

Product Code. 10092274

  • 50.80 € / Each
Estimated Shipment: 11-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Constructed in alloy and stainless steel
  • Accepts articles up to 125mm in diameter
  • Actuated by nickel-plated brass pummels and twin screw
  • Jaws are fitted with rubber sleeves for a soft grip

Specifications

Specifications

Prong Clamp
5 in.
Pivoted, Rubber Sleeves
Alloy, Stainless Steel
125 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.