missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProPac™ Elite WCX HPLC Column

This column provides consistent separations of therapeutic proteins including monoclonal antibodies, antibody drug conjugates, biosimilars and charge variants.

Merk:  Thermo Scientific™ 303355

3732.99 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 Bekijk meerdere types van dit product via de

Produktcode. 16575934

  • 3751.65 € / Each
Estimated Shipment: 27-05-2025
to see stock.
Explore more special offers
Toevoegen aan winkelmandje
This item is not returnable. View return policy

Beschrijving

Beschrijving

Discover the unique Thermo Scientific™ ProPac™ Elite WCX weak cation exchange (WCX) liquid chromatography column. Designed for protein characterization including therapeutic proteins, monoclonal antibodies (mAbs), antibody drug conjugates (ADCs) and biosimilar proteins, this column offers high resolution, high efficiency separation of proteins and their charge variants.

The unique column chemistry provides excellent performance under a broad range of pH, temperature and mobile phase compositions with excellent recovery and low carryover.

Product Highlights
  • Superior resolution power for proteins, monoclonal antibodies, and associated charge variants
  • High efficiency with reproducible separa-tions
  • High recovery with low carry-over
  • Wide pH operating range: 2 – 12
  • High temperature stability: up to 60°C
  • High throughput
  • Compatible with CX-1 pH Gradient Buffers
TRUSTED_SUSTAINABILITY
Specificatie

Specificatie

Analytical Column
150 mm
Recommended: Goods Buffers and NaCl. Minimum 20 mM salt required. Do not rinse with pure DI H2O
2 to 12
1 Column
Carboxylate
Nonporous DVB Particles with Hydrophilic Coating
Ion Exchange
Responsibly packaged
9 mm
4500 psi (310 bar)
5 μm
Nonporous
10°C to 60°C
ProPac
Ion Exchange
Bio-LC
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado