missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ProPac™ HIC-10 HPLC Columns

Achieve high-resolution separation of proteins and peptides under nondenaturing conditions with Thermo Scientific™ ProPac™ HIC-10 LC Columns.

Brand:  Thermo Scientific™ 063665

 View more versions of this product

Product Code. 11321863

  • 1522.63 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Achieve high-resolution separation of proteins and peptides under nondenaturing conditions with Thermo Scientific™ ProPac™ HIC-10 LC Columns. Developed with multi-functional ligand attachments, these silica-based columns provide exceptional hydrolytic stability under the highly aqueous conditions used in HIC.

High-efficiency Columns for the Separation of Large Biomolecules
  • High protein loading capacity for protein purification applications
  • Wide range of analytical and preparative applications, including the separation of bovine serum proteins, snake venom proteins, and monoclonal antibodies (MAbs) 
  • Based on 5μm ultra-high purity spherical silica gel particles with 300Å pores
Hydrophobic interaction chromatography separates biomolecules in a decreasing salt gradient, based on differences in surface hydrophobicity. This method preserves the biological activity of proteins. The HIC separation mechanism is complementary to those of ion-exchange and gel filtration chromatography.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Analytical Column
75 mm
5 μm
300 Å
40°C
ProPac
Hydrophobic Interaction
Stainless Steel
Bio-LC
7.8 mm
3500 psi (240 bar)
2.5 to 7.5
100 m²/g
Amide/Ethyl
Spherical, Fully Porous, Ultrapure Silica
Hydrophobic Interaction
Analytical Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.