missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propane-1,2-Diol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP, Fisher Chemical™
28.40 € - 485.00 €
Chemical Identifiers
CAS | 57-55-6 |
---|---|
Molecular Formula | C3H8O2 |
Molecular Weight (g/mol) | 76.095 |
MDL Number | 64272 |
InChI Key | DNIAPMSPPWPWGF-UHFFFAOYSA-N |
Synonym | 1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost |
PubChem CID | 1030 |
ChEBI | CHEBI:16997 |
IUPAC Name | propane-1,2-diol |
SMILES | CC(CO)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10346290
|
Fisher Chemical
P/7440/08 |
500 mL | Amber glass bottle |
28.40 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10518070
|
Fisher Chemical
P/7440/17 |
2.5 L | Amber glass bottle |
130.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10499370
|
Fisher Chemical
P/7440/25 |
25 L | Metal can |
485.00 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Propane-1,2-Diol, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, USP
Chemical Identifiers
57-55-6 | |
76.095 | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
1030 | |
propane-1,2-diol |
C3H8O2 | |
64272 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
CHEBI:16997 | |
CC(CO)O |
Specifications
57-55-6 | |
187°C | |
64272 | |
DNIAPMSPPWPWGF-UHFFFAOYSA-N | |
propane-1,2-diol | |
1030 | |
CHEBI:16997 | |
0.13mbar at 20°C |
-60°C | |
C3H8O2 | |
1,2-propanediol, propylene glycol, 1,2-dihydroxypropane, 2-hydroxypropanol, 1,2-propylene glycol, methylethyl glycol, methylethylene glycol, monopropylene glycol, isopropylene glycol, dowfrost | |
CC(CO)O | |
76.095 | |
2.62 | |
76.1g/mol | |
Viscous Liquid |