missing translation for 'onlineSavingsMsg'
Learn More
Propargyl alcohol, 99%, Thermo Scientific Chemicals Product Code.: 10001440

Propargyl alcohol, 99%, Thermo Scientific Chemicals

Product Code. 10001440
500 mL, Glass bottle
Click to view available options
Quantity:
250 mL
500 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10001440

Brand: Thermo Scientific Acros 131455000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-19-7, 50-00-0
Molecular Formula C3H4O
Molecular Weight (g/mol) 56.06
MDL Number MFCD00002912
InChI Key TVDSBUOJIPERQY-UHFFFAOYSA-N
Synonym propargyl alcohol, 2-propyn-1-ol, ethynylcarbinol, 1-propyn-3-ol, propynyl alcohol, 2-propynyl alcohol, 2-propynol, 3-propynol, 1-hydroxy-2-propyne, 3-hydroxy-1-propyne
PubChem CID 7859
ChEBI CHEBI:28905
IUPAC Name prop-2-yn-1-ol
SMILES OCC#C

Specifications

Melting Point -53.0°C
Color Colorless to Yellow
Density 0.9490g/mL
Boiling Point 114.0°C to 115.0°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2OH
Refractive Index 1.4300 to 1.4330
Quantity 500 mL
Beilstein 01,454
Fieser 09,394
Merck Index 15,7923
Specific Gravity 0.949
Solubility Information Solubility in water: miscible. Other solubilities: miscible with benzene, chloroform, ethanol, ether,, 1,2-dichloroethane, acetone, dioxane, thf,pyridine, moderately soluble in ccl4, immiscible with aliphatic hydrocarbons
Viscosity 1.58 mPa.s (20°C)
Formula Weight 56.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Propargyl alcohol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propargyl alcohol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Propargyl alcohol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.