missing translation for 'onlineSavingsMsg'
Learn More

Propargyl bromide, 80% in toluene, stab. with MgO, Thermo Scientific Chemicals

Product Code. 11477907
Click to view available options
Quantity:
50 mL
250 mL
1000 mL

Propargyl bromide, 80% in toluene is used to prepare betulonic acid-peptide conjugates with anti-inflammatory activity and propargylic amines employed in enyne metathesis. It finds application in the propargylation of spiro ketones, allylic alcohols and enone complexes. In Barbier-type reaction, it reacts with aldehydes to give alkyne alcohols. It is actively involved in the synthesis of polyethylene glycol (PEG) and peptide-grafted polyesters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl bromide, 80% in toluene is used to prepare betulonic acid-peptide conjugates with anti-inflammatory activity and propargylic amines employed in enyne metathesis. It finds application in the propargylation of spiro ketones, allylic alcohols and enone complexes. In Barbier-type reaction, it reacts with aldehydes to give alkyne alcohols. It is actively involved in the synthesis of polyethylene glycol (PEG) and peptide-grafted polyesters.

Solubility
Miscible with ethanol, ether, benzene, carbon tetrachloride and chloroform. Immiscible with water.

Notes
Light sensitive. Store in a cool place. Incompatible withbases, strong oxidizing agents, copper, peroxides, silver, silver oxides, mercury and mercury oxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-96-7
Molecular Formula C3H3Br
Molecular Weight (g/mol) 118.961
MDL Number MFCD00000241
InChI Key YORCIIVHUBAYBQ-UHFFFAOYSA-N
Synonym propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
PubChem CID 7842
IUPAC Name 3-bromoprop-1-yne
SMILES C#CCBr

Specifications

Melting Point -61°C
Density 1.335
Boiling Point 88°C to 90°C
Flash Point 3°C (37°F)
Refractive Index 1.494
Quantity 50 mL
UN Number UN2924
Beilstein 605309
Sensitivity Light sensitive
Solubility Information Miscible with ethanol,ether,benzene,carbon tetrachloride and chloroform. Immiscible with water.
Formula Weight 118.97
Concentration or Composition (by Analyte or Components) 80% in toluene
Chemical Name or Material Propargyl bromide, stabilized with MgO
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propargyl bromide, 80% in toluene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Propargyl bromide, Toluene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.