missing translation for 'onlineSavingsMsg'
Learn More

Propargyl bromide, 80wt.% solution in toluene, stabilized, AcroSeal™, Thermo Scientific Chemicals

106.53 € - 671.34 €

Specifications

Boiling Point 88.0°C to 90.0°C
Chemical Name or Material Propargyl bromide
Name Note Stabilized
CAS 106-96-7, 108-88-3
Color White to Yellow
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10166924
View Documents
Thermo Scientific Acros
429121000
100 mL AcroSeal™ Glass Bottle
106.53 €
100mL
Please to purchase this item. Need a web account? Register with us today!
15179293
View Documents
Thermo Scientific Acros
429120010
1 L AcroSeal™ Glass Bottle
671.34 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
Specifications

Specifications

88.0°C to 90.0°C
Stabilized
White to Yellow
C3H3Br
MFCD00000241
propargyl bromide, 3-bromopropyne, 3-bromo-1-propyne, 1-propyne, 3-bromo, 2-propynyl bromide, propynyl bromide, 1-bromo-2-propyne, propyne, 3-bromo, gamma-bromoallylene, 1-brom-2-propin
YORCIIVHUBAYBQ-UHFFFAOYSA-N
3-bromoprop-1-yne
7842
73 to 87% (propargyl bromide) (GC), 13 to 27% (toluene) (GC)
1.4900 to 1.4960
1.3800g/mL
Propargyl bromide
106-96-7, 108-88-3
Crystalline Powder
HC≡CCH2Br
4°C
Solubility in water: immiscible
C#CCBr
118.96
118.96
AcroSeal™ Glass Bottle
1.38
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure. Suspected of damaging fertility or the unborn child. May cause drowsiness or dizziness. May be fatal if swallowed and enters airways. Causes severe skin burns and eye damage. Toxic if swallowed. Highly flammable liquid and vapor.

GHS P Statement
Use personal protective equipment as required. Avoid breathing dust/fume/gas/mist/vapors/spray. Do not breathe dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. IF ON SKIN: Wash with plenty of soap and water. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-447-1

Product Identifier
  • Propargyl bromide, 80 wt% solution in toluene, stabilized

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only