missing translation for 'onlineSavingsMsg'
Learn More
Propargylamine, 98%, Thermo Scientific Chemicals Product Code.: 15470378

Propargylamine, 98%, Thermo Scientific Chemicals

Product Code. 15470378
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15470378

Brand: Thermo Scientific Alfa Aesar H53495.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Propargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargylamine is used in the synthesis of a chiral, a fluorescent macrocycle by 1,3-dipolar cycloaddition of propargyl amides of carbohydrate-linked amino acids and 9,10-bis(azidomethyl)anthracene. It acts as an intermediate in the production of drugs.

Solubility
Miscible with water, chloroform and ethyl acetate.

Notes
Hygroscopic. Light sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, strong oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2450-71-7
Molecular Formula C3H5N
Molecular Weight (g/mol) 55.08
MDL Number MFCD00008198
InChI Key JKANAVGODYYCQF-UHFFFAOYSA-N
Synonym propargylamine, 2-propynylamine, 2-propyn-1-amine, 3-aminopropyne, 3-amino-1-propyne, 2-propyn-1-thiol, 3-aminoprop-1-yne, propargylamin, pro-pargylamine, propargyl amine
PubChem CID 239041
IUPAC Name prop-2-yn-1-amine
SMILES C#CCN

Specifications

Density 0.86
Boiling Point 83°C
Flash Point 6°C (43°F)
Linear Formula HC≡CCH2NH2
Refractive Index 1.449
Quantity 5 g
UN Number UN3286
Beilstein 773681
Sensitivity Air Sensitive
Solubility Information Miscible with water,chloroform and ethyl acetate.
Formula Weight 55.08
Percent Purity 98%
Chemical Name or Material Propargylamine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW19K052 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog NumberH53495.14,H53495.06
Lot Number10252541 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog NumberH53495.14,H53495.06
Lot NumberU12K050 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog NumberH53495.14,H53495.06
Lot Number10237149 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberH53495.14,H53495.06
Lot Number10237476 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberH53495.14,H53495.06
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propargylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 2-Propyn-1-amine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Propargylamine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.