Learn More
Bochem™ Propeller Stirrer, 4 Vertical Blades
Mix media with low and middle viscosity. Bochem™ Propeller Stirrer, 4 Vertical Blades is constructed of 18/10 stainless steel.
Brand: Bochem™ 9056
This item is not returnable.
View return policy
Description
- 18/10 stainless steel construction.
- Middle RPM range.
- Mixes media with low and middle viscosity.
Specifications
Vertical Blade | |
Centrifuge | |
100 mm |
Stainless Steel (18/10) | |
300 mm |