missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Propellers for ServoDyne™ Electronic Mixers

Three-bladed propeller for general purpose mixing

Brand:  Cole-Parmer™ 04552-45

 View more versions of this product

Product Code. 11810130

  • 92.95 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Can be used as single or mounted several on a shaft for thorough agitation
  • Electro-polished

  • Marine-style propellers: used for general-purpose mixing
  • Turbine style propellers: used for airless mixing in deep containers

Specifications

Specifications

3-blade Propeller
0.78 cm
3.8 cm
ServoDyne™ electronic mixer
0.31 in.
1.5 in.
Ideal for general purpose mixing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers