missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Propidium iodide, 95% Product Code.: 11996691

Thermo Scientific Chemicals Propidium iodide, 95%

Product Code. 11996691
25 mg, Glass bottle
Click to view available options
Quantity:
25 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11996691

Brand: Thermo Scientific Chemicals 440300250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25535-16-4
Molecular Formula C27H34I2N4
Molecular Weight (g/mol) 668.39
InChI Key XJMOSONTPMZWPB-UHFFFAOYSA-M
Synonym propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide
PubChem CID 104981
ChEBI CHEBI:51240
IUPAC Name 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
SMILES CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]

Specifications

Melting Point 220.0°C to 225.0°C
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Quantity 25 mg
Solubility Information Solubility in water: soluble.
Formula Weight 668.39
Percent Purity 95%
Chemical Name or Material Propidium iodide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0442118 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number440300250,440300010,440301000
Lot NumberA0463182 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number440300250,440300010,440301000
Lot NumberA0460129 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number440300250,440300010,440301000
Lot NumberA0451635 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number440300250,440300010,440301000
Lot NumberA0402777 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number440300250,440300010,440301000
compliance-icons compliance-icons
Product Identifier
  • Propidium iodide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Propidium iodide, 95% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.