missing translation for 'onlineSavingsMsg'
Learn More

Propidium Iodide, 99%, For HPLC analysis, MP Biomedicals™

Product Code. p-4405827 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mg
0.1 g
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 11435392

Brand: MP Biomedicals 0219545825

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 25535-16-4
Molecular Formula C27H34I2N4
Molecular Weight (g/mol) 668.406
InChI Key XJMOSONTPMZWPB-UHFFFAOYSA-M
Synonym propidium iodide, propidium diiodide, 3,8-diamino-5-3-diethyl methyl ammonio propyl-6-phenylphenanthridin-5-ium iodide, unii-tp416o228t, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenylphenanthridinium diiodide, 3,8-diamino-5-3-diethylaminopropyl-6-phenylphenanthridinium iodide methiodide, phenanthridinium, 3,8-diamino-5-3-diethylmethylammonio propyl-6-phenyl-, diiodide
PubChem CID 104981
ChEBI CHEBI:51240
IUPAC Name 3-(3,8-diamino-6-phenylphenanthridin-5-ium-5-yl)propyl-diethyl-methylazanium;diiodide
SMILES CC[N+](C)(CC)CCC[N+]1=C2C=C(C=CC2=C3C=CC(=CC3=C1C4=CC=CC=C4)N)N.[I-].[I-]

Specifications

Color Red
Physical Form Flaky Powder
Quantity 25 mg
Formula Weight 668.4
Percent Purity 99%
Chemical Name or Material Propidium Iodide, For HPLC analysis
compliance-icons compliance-icons
Product Identifier
  • Propidium Iodide
Signal Word
  • Warning
Hazard Category
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.