missing translation for 'onlineSavingsMsg'
Learn More
Propiolic Acid, 98%, Thermo Scientific Chemicals Product Code.: 10153470

Propiolic Acid, 98%, Thermo Scientific Chemicals

Product Code. 10153470
10 mL, Glass bottle
Click to view available options
Quantity:
10 mL
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10153470

Brand: Thermo Scientific Acros 131500100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-25-0
Molecular Formula C3H2O2
Molecular Weight (g/mol) 70.05
MDL Number MFCD00004360
InChI Key UORVCLMRJXCDCP-UHFFFAOYSA-N
Synonym propiolic acid, 2-propynoic acid, propynoic acid, propargylic acid, acetylenecarboxylic acid, carboxyacetylene, propiolic acidd, propiolicacid, unii-p2qw39g9lz, propynoate
PubChem CID 10110
ChEBI CHEBI:33199
IUPAC Name prop-2-ynoic acid
SMILES OC(=O)C#C

Specifications

Melting Point 16.0°C
Color Colorless to Brown
Density 1.1300g/mL
Boiling Point 153.0°C to 157.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HC≡CCO2H
Refractive Index 1.4315 to 1.4335
Quantity 10 mL
Merck Index 15, 7935
Specific Gravity 1.13
Solubility Information Solubility in water: miscible
Formula Weight 70.04
Percent Purity 98%
Physical Form Liquid After Melting
Chemical Name or Material Propiolic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466141 Certificate TypeCertificate of Analysis Date13/09/2024 Catalog Number131500250,131500100,131501000,13150016
Lot NumberA0462239 Certificate TypeCertificate of Analysis Date29/04/2024 Catalog Number131500250,131500100,131501000,13150016
Lot NumberA0457898 Certificate TypeCertificate of Analysis Date30/11/2023 Catalog Number131500250,131500100,131501000,13150016
Lot NumberA0399089 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number131500250,131500100,131501000,13150016
Lot NumberA0414959 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number131500250,131500100,131501000,13150016
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Propiolic acid
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Propiolic Acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.