missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionic acid, ≥99.5% (GC), Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
50 mL
Packaging:
Poly bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
CAS | 79-09-4 |
Melting Point | -21°C |
Color | Colorless |
Density | 0.993g/mL at 25°C |
pH | Acidic |
Boiling Point | ∼141°C (1013hPa) |
Flash Point | 54°C (129.2°F) |
Ignition Point | 955°F |
Odor | Unpleasant |
Packaging | Poly bottle |
Show More |
Safety and Handling



Product Identifier
- Propionic acid
- Danger
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Propionic acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction