missing translation for 'onlineSavingsMsg'
Learn More

Propionic anhydride, 99

Product Code. 10669783 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Unit Size:
1 L
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10669783

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

CAS: 123-62-6 | C6;H1;0;O3; | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-62-6
Molecular Formula C6;H1;0;O3;
Molecular Weight (g/mol) 130.14

Specifications

Melting Point -42.0°C
Density 1.0150 g/mL
Boiling Point 167.0°C to 170.0°C
Flash Point 63°C
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4030 to 1.4050
Quantity 250 mL
Beilstein 02, 242
Merck Index 15, 7940
Specific Gravity 1.0150
Solubility Information Solubility aq. soln.: hydrolyses aq. soln. forming propionic acid. Other solubilities: soluble in methanol, ethanol, ether and chloroform
Viscosity 1.144 mPa.s (20°C)
Formula Weight 130.14
Chemical Name or Material Propionic anhydride
Show More Show Less
compliance-icons
Product Identifier
  • Propionic anhydride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.