missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propiophenone, 99%, Thermo Scientific Chemicals
23.30 € - 175.00 €
Chemical Identifiers
CAS | 93-55-0 |
---|---|
Molecular Formula | C9H10O |
Molecular Weight (g/mol) | 134.18 |
InChI Key | KRIOVPPHQSLHCZ-UHFFFAOYSA-N |
Synonym | propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one |
PubChem CID | 7148 |
ChEBI | CHEBI:425902 |
IUPAC Name | 1-phenylpropan-1-one |
SMILES | CCC(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10715791
|
Thermo Scientific Acros
131540020 |
2 mL | Glass bottle | ||||||
10439060
|
Thermo Scientific Acros
131542500 |
250 mL | Glass bottle |
29.30 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10051470
|
Thermo Scientific Acros
131540010 |
1 L | Glass bottle |
85.10 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11934851
|
Thermo Scientific Acros
131540025 |
2.5 L | Glass bottle |
175.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-55-0 | |
134.18 | |
propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one | |
CHEBI:425902 | |
CCC(=O)C1=CC=CC=C1 |
C9H10O | |
KRIOVPPHQSLHCZ-UHFFFAOYSA-N | |
7148 | |
1-phenylpropan-1-one |
Specifications
93-55-0 | |
Colorless to Yellow | |
218.0°C | |
Authentic | |
C9H10O | |
1.5248 to 1.5268 | |
15,7944 | |
propiophenone, ethyl phenyl ketone, propionylbenzene, 1-phenyl-1-propanone, 1-propanone, 1-phenyl, phenyl ethyl ketone, ketone, ethyl phenyl, propionphenone, usaf ek-1235, 1-phenyl-propan-1-one | |
KRIOVPPHQSLHCZ-UHFFFAOYSA-N | |
1-phenylpropan-1-one | |
7148 | |
134.18 | |
Liquid |
18.0°C | |
1.0000g/mL | |
87°C | |
98.5% min. (GC) | |
C6H5COC2H5 | |
07,II,231 | |
1 | |
Solubility in water: insoluble. Other solubilities: miscible with methanol, ether, benzene and toluene, insoluble in glycerol, ethylene glycol and, propylene glycol | |
CCC(=O)C1=CC=CC=C1 | |
134.18 | |
CHEBI:425902 | |
99% | |
Propiophenone |
Safety and Handling
missing translation for 'einecsNumber' : 202-257-6
RUO – Research Use Only