missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Propranolol hydrochloride, 99%, Thermo Scientific Chemicals

75.27 € - 238.19 €

Chemical Identifiers

CAS 318-98-9
Molecular Formula C16H22ClNO2
Molecular Weight (g/mol) 295.807
MDL Number MFCD00012558
InChI Key ZMRUPTIKESYGQW-UHFFFAOYSA-N
Synonym propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal
PubChem CID 62882
ChEBI CHEBI:8500
IUPAC Name 1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride
SMILES CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11372539
View Documents
Thermo Scientific Alfa Aesar
H26645.06
5 g
75.27 €
5g
Please to purchase this item. Need a web account? Register with us today!
11382539
View Documents
Thermo Scientific Alfa Aesar
H26645.14
25 g
238.19 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

A beta-adrenergic blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

318-98-9
295.807
ZMRUPTIKESYGQW-UHFFFAOYSA-N
62882
1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride
C16H22ClNO2
MFCD00012558
propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal
CHEBI:8500
CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl

Specifications

318-98-9
C16H22ClNO2
14,7840
ZMRUPTIKESYGQW-UHFFFAOYSA-N
1-naphthalen-1-yloxy-3-(propan-2-ylamino)propan-2-ol;hydrochloride
62882
295.8
(+/-)-Propranolol hydrochloride
163°C to 165°C
MFCD00012558
propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal
CC(C)NCC(COC1=CC=CC2=CC=CC=C21)O.Cl
295.807
CHEBI:8500
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

missing translation for 'einecsNumber' : 206-268-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • (+/-)-Propranolol hydrochloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4

Hazard Statement
  • H302-Harmful if swallowed.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : [2-Hydroxy-3-(naphthyloxy)propyl]isopropyl ammonium chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.