missing translation for 'onlineSavingsMsg'
Learn More

Propranolol hydrochloride, Tocris Bioscience™

Product Code. 30001108
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

β antagonist

β-adrenergic antagonist.

R-enantiomer, S-enantiomer and Glycol Metabolite also available.

Specifications

CAS 3506-09-0
Molecular Formula C17H24ClNO2
MDL Number MFCD00012558
Quantity 100 mg
Synonym propranolol hydrochloride, propranolol hcl, inderal, avlocardyl, herzbase, inderalici, naprilin, pronovan, dociton, ikopal
InChI Key RTTYPFXTLKDXPS-UHFFFAOYNA-N
SMILES [H+].[Cl-].CC(C)CNCC(O)COC1=C2C=CC=CC2=CC=C1
IUPAC Name hydrogen 1-[(2-methylpropyl)amino]-3-(naphthalen-1-yloxy)propan-2-ol chloride
Molecular Weight (g/mol) 309.83
PubChem CID 62882
ChEBI CHEBI:8500
Formula Weight 295.81
Percent Purity >98%
Chemical Name or Material Propranolol hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'