missing translation for 'onlineSavingsMsg'
Learn More

Propylene carbonate, CHROMASOLV™, for HPLC, 99.7%, Honeywell Riedel-de Haën™

Product Code. 15685450
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CHROMASOLV™, for HPLC, 99.7%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

CAS 108-32-7
CAS Max % ≤100.0000%
Melting Point -55°C (literature)
Density 1.204 g/mL at 25 °C(lit.)
Boiling Point 242°C
Flash Point 135°C
Packaging Glass Bottle
Molecular Formula C4H6O3
Linear Formula C4H6O3
MDL Number MFCD00005385,MFCD00798264,MFCD00798265
Quantity 1 L
Beilstein 107913
Synonym propylene carbonate, 1,2-propylene carbonate, 1,2-propanediol cyclic carbonate, texacar pc, 1,3-dioxolan-2-one, 4-methyl, cyclic propylene carbonate, arconate 5000, 1,2-propanediol carbonate, 1-methylethylene carbonate, cyclic 1,2-propylene carbonate
InChI Key RUOJZAUFBMNUDX-UHFFFAOYNA-N
SMILES CC1COC(=O)O1
IUPAC Name 4-methyl-1,3-dioxolan-2-one
Molecular Weight (g/mol) 102.09
PubChem CID 7924
Percent Purity >99.65%
Grade HPLC
Chemical Name or Material Propylene carbonate, CHROMASOLV™, for HPLC, 99.7%
Mehr anzeigen Weniger anzeigen
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'