missing translation for 'onlineSavingsMsg'
Learn More

Propylenediphosphonic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11397238
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4671-82-3
Molecular Formula C3H10O6P2
Molecular Weight (g/mol) 204.055
MDL Number MFCD00050066
InChI Key DZQCAOQMXPROIJ-UHFFFAOYSA-N
Synonym propylenediphosphonic acid, acmc-20ajta, propylene-bisphosphonic acid, propylenediphosphonicacid, propane-1,2-diylbis phosphonic acid, 1-phosphonopropan-2-yl phosphonic acid
PubChem CID 22023450
IUPAC Name 1-phosphonopropan-2-ylphosphonic acid
SMILES CC(CP(=O)(O)O)P(=O)(O)O

Specifications

Melting Point 178°C
Odor Odorless
Quantity 0.5 g
Formula Weight 204.06
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Propylenediphosphonic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'