Learn More
Contec™ PROSAT™ Pi Microfiber Wipes
Microfiber wipes presaturated with 70% IPA and 30% DI water.
Brand: Contec™ PS-314IR-BPR
738.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
PROSAT™ Pi wipes are designed to help life science manufacturers control contamination and ultimately maintain cGMP compliance while consuming less resources. The wipes are made from polyester and nylon split microfiber, enhancing their cleaning effectiveness and contaminant pick-up ability. The 80 gsm microfiber fabric is highly sorbent and offers metered release of solvent to surfaces, covering greater areas compared to other presaturated wipes.
PROSAT Pi wipes are presaturated with 70% USP grade IPA and 30% DI water, allowing for controlled application to surfaces and less VOCs compared to spraying. The peel-and-reseal pouch can be resealed throughout use to maintain the wipes’ saturation level and IPA concentration.
PROSAT Pi wipes are ideal for pharmaceutical, biotechnology and medical device manufacturers.
- Nonwoven split microfiber increases cleaning effectiveness and contaminant pick up and offers a smooth, even laydown of solvent to surfaces
- Covers greater surface area compared to non-microfiber wipes, reducing the number of wipes required for some cleaning tasks
- Presaturated wipes have controlled saturation levels greatly reduce solvent usage, VOCs and waste
Specifications
Not Autoclavable | |
Yes | |
23 x 23 cm (9 x 9 in.) | |
70% IPA / 30% DI Water |
White | |
Guillotine | |
Work Benches, Tables, etc. | |
Sterile |
Safety and Handling
- X800 PROSAT Sterile, Nonwoven, microfiber wipes wi
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.