Learn More
Contec™ PROSAT™ Presaturated Nonwoven Wipes



Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
16.00 € - 969.00 €
Specifications
Product Type | Pre-saturated Wipe |
---|---|
Autoclavable | Not Autoclavable |
Disposable | Single-use |
Ply | 1-ply |
Sterility | Non-sterile |
Product Code | Brand | Dimensions (L x W) | Material | Saturation Solution | Packaging Quantity | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Material | Saturation Solution | Packaging Quantity | Price | Quantity & Availability | ||
15755533
![]() ![]() |
Contec™
PSPP0043 |
28 x 23 cm (11 x 9 in.) | Polypropylene | 70% IPA, 30% DI Water | 50 Wipes/Bag, 40 Bags/Cs. | N/A | |||
15823424
![]() |
Contec™
PSPP0039 |
28 x 23 cm (11 x 9 in.) | Polypropylene | 70% IPA, 30% DI Water | 1800 Wipes/Cs. | N/A | |||
17244513
![]() |
Contec™
PS-850-BPR |
20 x 20 cm (8 x 8 in.) | Polypropylene | 70% IPA, 30% DI Water | 50 Pk., 50 Pk./Cs. |
774.00 €
Pack of 2500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11952435
![]() |
Contec™
PS-850 |
20 x 20 cm (8 x 8 in.) | Polypropylene | 70% IPA, 30% DI Water | 50 Wipes/Bag | N/A | |||
15626996
![]() |
Contec™
PS-850 |
20 x 20 cm (8 x 8 in.) | Polypropylene | 70% IPA, 30% DI Water | 50 Wipes/Bag | N/A | |||
17254513
|
Contec™
PS-850E-BPR |
20 x 20 cm (8 x 8 in.) | Polypropylene | 70% IPA, 30% DI Water | 50 Wipes/Bag, 40 Bags/Cs. | N/A | |||
17234523
![]() |
Contec™
PSC20002-BPR |
28 x 23 cm (11 x 9 in.) | Polyester Cellulose | 70% IPA, 30% DI Water | 24 Pk., 50 Pk./Cs. |
923.00 €
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17254523
![]() |
Contec™
PSC20006-BPR |
28 x 23 cm (11 x 9 in.) | Polyester Cellulose | 70% IPA, 30% DI Water | 50 Pk., 28 Pk./Cs. |
764.00 €
Pack of 1400 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
17204533
![]() |
Contec™
PSPP0039-BPR |
28 x 23 cm (11 x 9 in.) | Polypropylene | 70% IPA, 30% DI Water | 50 Pk., 24 Pk./Cs. |
875.00 €
Pack of 1200 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
12817163
![]() |
Contec™
PSC20002 |
28 x 23 cm (11 x 9 in.) | Polyester Cellulose | 70% IPA, 30% DI Water | 24 Wipes/Bag | N/A | |||
11962435
![]() |
Contec™
PSC20006 |
28 x 23 cm (11 x 9 in.) | Polyester Cellulose | 70% IPA, 30% DI Water | 50 Wipes/Bag | N/A | |||
17214533
![]() |
Contec™
PSPP0043-BPR |
28 x 23 cm (11 x 9 in.) | Polypropylene | 70% IPA, 30% DI Water | 50 Pk., 40 Pk./Cs. |
969.00 €
Pack of 2000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. Contec™ PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
- Wipe substrate designed for the application
- Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
- Ideal for wiping articles prior to pass through
- Made of meltblown polypropylene or cellulose/polyester
- Effective and convenient for wipe-downs of process tools and other equipment
- Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipes for improved durability, or economical hydroentangled polyester/cellulose wipers
- Solvent blend meets SEMI specifications for metals and contaminants
Specifications
Pre-saturated Wipe | |
Single-use | |
Non-sterile |
Not Autoclavable | |
1-ply |
Safety and Handling
- CONTEC PROSAT MBPP 70/30 IPA & DI WATER
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use media other than water to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol
Your input is important to us. Please complete this form to provide feedback related to the content on this product.