missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ QuickTask Mop - Presaturated with 70% IPA, 30% DI Water
GREENER_CHOICE

Product Code. p-7250216 Shop All Contec Products
Click to view available options
Model:
PSTA0021
PSTA0022, Irradiated
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 30
This item is not returnable. View return policy

Product Code. 17800194

Brand: Contec™ PSTA0022BPR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mops presaturated with USP Grade IPA provide consistency between operators and eliminate the need to take extra cleaning solution and separate mops and buckets into a room. Presaturated mops allow for targeted application of IPA to surfaces. They eliminate the risk of flammable liquid spills and reduce IPA usage and VOCs.

The PROSAT™ QuickTask™ mopping system provides a consistent and convenient option when using IPA to clean or remove residues. Each mop is presaturated with a consistent amount of 70% USP grade isopropyl alcohol, eliminating the need for additional accessories such as bucket systems or wringers. These mops are packaged in a convenient easy to tear pouch. The system has a 100% polyester face and generates low levels of particles and fibers. Using this mop with the QuickTask™ frame and QuickConnect™ handle makes it an ergonomic alternative to hand wiping with IPA presaturated wipes. This system is ideal for rinsing after applying disinfectants to ceilings, walls, and floors, or for removing dried-on residues from surfaces. The PROSAT QuickTask™ mopping system is available irradiated for use in Grade B to D cleanrooms.

IPA presaturated QuickTask™ mop

  • 100% 2-ply polyester covered mop heads release low levels of particles and fibers
  • Available irradiated, making it ideal for use in Grade B cleanrooms
  • Packaging is a convenient easy to tear pouch
  • Small number of mops per package limits excess waste, making it ideal for smaller environments

Packaging:
Packaged in polyethylene outer bags to simplify product transfer.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Mop
Model PSTA0022, Irradiated
Length (English) 16 in.
Sterility Sterile
Length (Metric) 41 cm
Autoclavable Not Autoclavable
Width (Metric) 13 cm
Width (English) 5 in.
Disposable Single-use
Color-coded No
Material Polyester
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Contec IPA Wipes
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P370+P378-In case of fire: Use alcohol resistant foam, water fog to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Propan-2-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.