missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec PROSAT ReFIBE Wipes
Presaturated wipes manufactured from 100% recyled knitted polyester cleanroom wipes.
Brand: Contec PSRF99-BPR
334.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- 100% knitted laundered recycled polyester provides low levels of particles and fibers
- Abrasion and chemical resistant
- Soft textured wipe that cleans sufaces without scratching them
- Manufactured from 100% recycled plastic diverts plastic from oceans and landfill
- Helps organization reduce its environmental impact and meet its sustainability goals
- Presaturated wipes reduce solvent usage and VOC emissions
- Ensures consistent saturation of each wipe independent of the operator
- Reseable pouch maintains satuation levels throughout use and makes wipe removal easy
- Laser cut sealed edges seal fibers at wipes edge reducing shedding
- Validated sterile to a SAL of 10-6 suitable for use in the highest grade cleanrooms
Specifications
Wipes | |
230 mm | |
230 mm | |
Not Autoclavable | |
Interlock knit, no-run | |
Non-sterile |
9 in. | |
9 in. | |
Polyester | |
124 gsm | |
Single-use |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction