missing translation for 'onlineSavingsMsg'
Learn More

Contec PROSAT ReFIBE Wipes
GREENER_CHOICE

Presaturated wipes manufactured from 100% recyled knitted polyester cleanroom wipes.

Brand:  Contec PSRF1212IR-BPR

644.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 17856693

  • 995.00 € / Pack of 400
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 100% knitted laundered recycled polyester provides low levels of particles and fibers
  • Abrasion and chemical resistant
  • Soft textured wipe that cleans sufaces without scratching them
  • Manufactured from 100% recycled plastic diverts plastic from oceans and landfill
  • Helps organization reduce its environmental impact and meet its sustainability goals
  • Presaturated wipes reduce solvent usage and VOC emissions
  • Ensures consistent saturation of each wipe independent of the operator
  • Reseable pouch maintains satuation levels throughout use and makes wipe removal easy
  • Laser cut sealed edges seal fibers at wipes edge reducing shedding
  • Validated sterile to a SAL of 10-6 suitable for use in the highest grade cleanrooms
Specifications

Specifications

Wipes
300 mm
300 mm
Not Autoclavable
Interlock knit, no-run
Sterile
12 in.
12 in.
Polyester
124 gsm
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers