missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ PROSAT™ Sterile Meltblown Polypropylene LE Wipes
Contec™ PROSAT™ Sterile Meltblown Polypropylene LE Wipes provide a consistent release of solvent to thoroughly remove surface contaminants in critical environments.
Brand: Contec™ PS-911LE-BPR
This item is not returnable.
View return policy
Description
- Suitable for Grade A or ISO Class 5 environments
- Validated sterile wipe certified to a guaranteed low endotoxin level of <20 EU/wipe so ideal for product contact areas
- Low in particles and fibres so ideal for a variety of critical applications
Specifications
Pre-saturated Wipe | |
Grade A/B or ISO Class 5 environments | |
23 x 28 cm | |
Sterile |
Meltblown Polypropylene | |
Single-use | |
30 Pk., 36 Pk./Cs. |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P411-Store at temperatures not exceeding °C/°F.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction