missing translation for 'onlineSavingsMsg'
Learn More

Contec™ PROSAT™ Sterile Meltblown Polypropylene LE Wipes
BIOPHARMAGREENER_CHOICE

Contec™ PROSAT™ Sterile Meltblown Polypropylene LE Wipes provide a consistent release of solvent to thoroughly remove surface contaminants in critical environments.

Brand:  Contec™ PS-911LE


Product Code. 15762667

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Suitable for Grade A or ISO Class 5 environments
  • Validated sterile wipe certified to a guaranteed low endotoxin level of <20 EU/wipe so ideal for product contact areas
  • Low in particles and fibres so ideal for a variety of critical applications
Specifications

Specifications

Pre-saturated Wipe
Grade A/B or ISO Class 5 environments
23 x 28 cm
Sterile
Meltblown Polypropylene
Single-use
30/Pouch, 36 Pouches/Cs.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • contec prosat MBPP with 70% IPA

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : propan-2-ol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers