missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ ProShield™ 60
ProShield™ 60 provides a barrier against non-hazardous dry particles down to one micron in size.
340.00 €
Specifications
Gender | Unisex |
---|---|
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
16234511
|
DuPont™
D15519552 |
Small |
340.00 €
Pack of 50 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
16244511
|
DuPont™
D15519553 |
Medium |
340.00 €
Pack of 50 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
16254511
|
DuPont™
D15519554 |
Large |
340.00 €
Pack of 50 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
16264511
|
DuPont™
D15519555 |
X-Large |
340.00 €
Pack of 50 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
16274511
|
DuPont™
D15519556 |
2X-Large |
340.00 €
Pack of 50 |
Estimated Shipment: 10-10-2024
Log in to see stock. |
|||||
Description
- Constructed of a microporous film on a polypropylene nonwoven fabric.
- Combine good particle protection with high liquid repellency.
- Help provide a barrier against non-hazardous dry particles down to one micron in size.
- The material is permeable to moisture vapour only and is not air permeable.
- Certified according to Regulation (EU) 2016/425.
- Chemical protective clothing, Category III, Type 5 and 6.
- EN 1073-2 (protection against radioactive contamination).
- Antistatic treatment (EN 1149-5) on inside.
- Stitched internal seams.
- Nylon zipper with flap.
- Ideal choice for applications that are less demanding in terms of barrier, durability and comfort.
Specifications
Unisex | |
White | |
Elastic | |
Elastic | |
Zipper with Storm Flap | |
Non-sterile | |
50/Cs. |
Polypropylene | |
Single-use | |
Elastic | |
Hooded | |
Stitched | |
Individually Packed |