missing translation for 'onlineSavingsMsg'
Learn More
DuPont™ ProShield™ 20 Blue Product Code.: 15421255
Estimated Shipment: 13-05-2025
to see stock.
Add to basket

DuPont™ ProShield™ 20 Blue

Product Code. 15421255 Shop All DuPont Products
Small
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy

Product Code. 15421255

Brand: DuPont™ D15338174

Estimated Shipment: 13-05-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to protect workers from certain substances with a high level of comfort. Dupont™ ProShield™ 20 Blue Hooded Coverall is typically used for protection against particles (Type 5) and limited liquid splashes or sprays (Type 6).

  • Chemical protective clothing, Category III, Type 5 and 6
  • EN 1073-2 (protection against radioactive contamination)
  • Antistatic treatment (EN 1149-5) - on inside
  • Stitched external seams
  • Nylon zipper with flap
  • Available in blue in sizes S to XXXL (a white garment is also available).
  • 2-piece hood.
  • Elasticated face, wrists, waist and ankles
TRUSTED_SUSTAINABILITY

Specifications

Product Type Hooded Coveralls
Size Small
Material SMS Fabric
Color Blue
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Closure Type Zipper with Storm Flap
Seam Type Stitched
Protection Class Category III, Type 5 and 6
Certifications/Compliance (EU) 2016/425, EN 1073-2, EN 1149-5
Size (Metric) Chest 84 to 92 cm
For Use With (Application) For workers seeking protection against dirt and grime during light duty work and other industries
Quantity 50/Pk.
Show More Show Less

The user should make the risk assessment to determine the protection required.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ ProShield™ 20 Blue >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.