Learn More
PROTAC ERα Degrader-2, MedChemExpress
MedChemExpress PROTAC ERα Degrader-2 comprises a IAP ligand binding group, a linker and an estrogen receptor α (ERα) binding group. PROTAC ERα Degrader-2 is an ERα degrader. Maximal ERα degradation at 30 μM concentration in human mammary tumor MCF7 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs).
1035.45 € - 5578.20 €
Specifications
CAS | 1351169-29-3 |
---|---|
Molecular Formula | C42H61N5O8 |
Molecular Weight (g/mol) | 763.96 |
Percent Purity | 98.02% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16474538
|
Medchem Express
HY-111846 |
10 mg |
1792.80 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16427257
|
Medchem Express
HY-111846 |
5 mg |
1035.45 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16410847
|
Medchem Express
HY-111846 |
50 mg |
5578.20 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PROTAC ERα Degrader-2 comprises a IAP ligand binding group, a linker and an estrogen receptor α (ERα) binding group. PROTAC ERα Degrader-2 is an ERα degrader. Maximal ERα degradation at 30 μM concentration in human mammary tumor MCF7 cells. Degradation inducers based on cIAP1 are called specific and non-genetic IAP-dependent protein erasers (SNIPERs).Specifications
1351169-29-3 | |
C42H61N5O8 | |
763.96 | |
98.02% |
Safety and Handling
missing translation for 'shelfLife' : -20°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Recommended Storage : -20°C, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (stored under nitrogen)
Research purposes only