Learn More
Thermo Scientific Chemicals Protease Inhibitor Cocktail II
Brand: Thermo Scientific Chemicals J64963.LQ
104.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Inhibits aspartic, cysteine, serine, and metalloproteases, as well as aminopeptidases. Recommended for use with bacterial cell extracts
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Inhibits aspartic, cysteine, serine, and metalloproteases, as well as aminopeptidases
- Recommended for use with bacterial cell extracts
Specifications
White | |
Air sensitive | |
Lyophilized | |
1 Unit |
Protease Inhibitor Cocktail | |
-30°C to -10°C | |
Inhibits aspartic, cysteine, serine, and metalloproteases, as well as aminopeptidases. Recommended for use with bacterial cell extracts |
Safety and Handling
- Protease Inhibitor Cocktail II
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : N,N-1,2-ethanediylbis[N-(carboxymethyl)-glycine disodium salt, dihydrate, 4-(2-aminoethyl)-benzenes
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only