missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uvex™ Protection Flash Hi-Viz Orange Vest
Click to view available options
Size: Large
Description
- Vertical light guide system
- Hook-and-loop fastening to quickly close the vest
- Certified to EN ISO 20471 Class 2
- For use with light guide system: battery pack, art. no. 1733200
- Active warning
- With switch for easy operation
- Fast closure
- Easy to use
- Long run-time
- Areas of application: road construction works, structural and civil engineering, in dark environments
- Washing instructions: 40°C gentle cycle, do not use chlorine, do not tumble dry, do not iron, dry clean with perchlorethylene
Specifications
Specifications
Certifications/Compliance | EN ISO 20471, Class 2 |
Color | Orange |
Size | Large |
Material | Polyester |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'