Learn More
Cytiva Protein A
Brand:  Cytiva 17-0872-50
Product Code. 11349931
  • 951.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Staphylococcal Protein A is the classical immunoglobulin-binding reagent for a wide range of antibodies, used extensively in affinity chromatography and antibody detection applications. The protein is enterotoxin-free and gives reliable results.
  • Staphylococcal Protein A
  • Protein A binds the Fc regions of IgG antibodies from a variety of species, for convenient antibody purification.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Protein A