missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Protein A, Alexa Fluor™ 488 conjugate
Protein A, Alexa Fluor™ 488 conjugate
Brand: Molecular Probes™ P11047
212.04 EUR valid until 2024-09-27
Use promo code "21650" to get your promotional price.
This item is not returnable.
View return policy
Description
Our Alexa Fluor 488 protein A conjugate binds to the Fc portion of various immunoglobulins from several different species. This exceptionally bright, green-fluorescent probe (absorption/emission maxima ∼495/519 nm) can be used to visualize antibodies in a manner analogous to a labeled species-specific secondary antibody.Specifications
Alexa Fluor 488 | |
Alexa Fluor™ | |
Protein A | |
Room Temperature |
1 mg | |
Lyophilized Powder | |
Store in freezer (-5 to -30°C) and protect from light. |
For Research Use Only. Not for use in diagnostic procedures.