missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Protein A, Alexa Fluor™ 647 conjugate

Product Code. 10257192
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 10257192

Brand: Invitrogen™ P21462

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein A, Alexa Fluor™ 647 conjugate

Our Alexa Fluor™ 647 protein A conjugate binds to the Fc portion of various immunoglobulins from several different species. This exceptionally bright, far red-fluorescent probe (absorption/emission maxima ∼650/668 nm) can be used to visualize antibodies in a manner analogous to a labeled species-specific secondary antibody.
TRUSTED_SUSTAINABILITY

Specifications

Conjugate Alexa Fluor 647
Quantity 1 mg
Product Line Alexa Fluor
Form Lyophilized Powder
Protein Subtype Protein A
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.