missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein A/G HP SpinTrap™ Buffer Kit

Pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation using Protein A HP, Protein G HP, and Ab SpinTrap™ columns

Brand:  Cytiva 28-9135-67

307.80 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.



Product Code. 10545735

  • 324.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Sufficient quantities of the following reagents to perform 16 reactions: 10X Binding/Washing buffer, Classic elution buffer, 10X Crosslink elution buffer, 10X Crosslink Solution A, and 10X Crosslink Solution B.
This item is not returnable. View return policy

Description

Description

  • Eliminates time-consuming buffer preparation; allows fast, reproducible enrichment of target protein from complex protein mix
  • Contains reagents for both the Classic and Crosslink protocols (with/without elution of antibody together with target protein, respectively)
  • Convenient format: stock solutions can be diluted directly into the provided bottles for storage

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Protein A/G HP SpinTrap Buffer Kit
Pre-made buffers optimized for the affinity-based enrichment of a target protein/immunoprecipitation using Protein A HP, Protein G HP, and Ab SpinTrap columns.
4°C to 30°C
1 Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.