missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein A HP MultiTrap™ Plates

For small-scale purification of antibodies or enrichment of proteins of interest by antibody capture in 96-well filter plates

Brand:  Cytiva 28-9031-33


Product Code. 11733329

  • 2133.00 € / Pack of 4
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket

Requires : Four prepacked Protein A HP MultiTrap 96-well filter plates and instructions
This item is not returnable. View return policy

Description

Description

  • Flexible format, with protocols for both classic elution and cross-link methods
  • Formatted for elution for both electrophoresis and LC-MS analysis workflows
  • Fast-binding kinetics and high capacity provide high yield
  • Prepacked with Protein A Sepharose™ High Performance Medium for coupling of antibodies of IgG subclasses
  • Can be used with centrifugation or vacuum, manually or in automated robotic systems
  • Easy scale-up with HiTrap™ Protein A HP prepacked columns

  • Matrix: Highly cross-linked agarose, 6%
  • Binding capacity: approx. 20mg human IgG/mL medium
  • Average particle size: 34μm
  • pH stability: 3 to 9 (long term); 2 to 9 (short term)
  • Well volume: 800μL
  • Centrifugation speed: 700xG
  • Vacuum pressure: -0.5 bar (max.)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

20 mg/mL Human IgG Medium
70 to 130 μL
2°C to 8°C, 20% Ethanol
Centrifugation, Vacuum
For vacuum, centrifugation, manually or with robotic systems
4
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • protein A HP multitrap

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers