missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein A Mag Sepharose™ Xtra

Magnetic beads are designed for rapid, small-scale purification and screening of monoclonal and polyclonal antibodies from serum and cell supernatants

Brand:  Cytiva 28-9670-62

265.05 EUR valid until 2024-12-31
Use promo code "21631" to get your promotional price.



 View more versions of this product

Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10565210

  • 287.00 € / 5mL
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Used as a separation tool for handling the beads in microcentrifuge tubes, up to six samples can be processed in parallel and can be easily screen a larger number of samples in parallel with high throughput on a robotic device.

  • Visible and dense Sepharose-based magnetic beads with protein A ligand allow easy spotting and collection of bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation
  • Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Provides high purity and yield

∽350μg purified human IgG can be obtained from each purification run
Provides robust parallel screening of antibodies with high reproducibility
Matrix: paramagnetic, spherical, highly cross-linked agarose particles
Ligand: native protein A
Binding capacity: 27mg human IgG/mL gel
Operating temp.: room temperature
Stability: stable

  • Small-scale purification
  • Antibody screening
  • Antibody purification

TRUSTED_SUSTAINABILITY
Specifications

Specifications

27mg human IgG/mL gel
5 x 1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Protein A Mag Sepharose Xtra, 5 x 1 ml

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN ACUTE-66.5 per cent of the mixture consists of component(s) of unknown acute toxicity.
  • UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers