missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Protein A and Protein G Mag Sepharose™

Simplify enrichment of target proteins by immunoprecipitation or pull-down applications

62.41 € - 154.00 €

Specifications

Matrix Paramagnetic spherical, highly cross linked agarose particles
Particle Size 37 to 100 μm
Content And Storage 4°C to 8°C
View More Specs

Products 4
Product Code Brand Quantity Binding Capacity Ligand Bead Type Price Quantity & Availability  
Product Code Brand Quantity Binding Capacity Ligand Bead Type Price Quantity & Availability  
10300695
View Documents
Cytiva
28-9440-06
500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose
64.63 €
500µL
In Stock
to see stock.
Add to basket
 
10249574
View Documents
Cytiva
28-9440-08
500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose
62.41 €
500µL
In Stock
to see stock.
Add to basket
 
10488634
View Documents
Cytiva
28-9513-79
4 x 500 μL 13 to 22mg Human IgG/mL Protein G Protein G Mag Sepharose
134.85 €
Pack of 4
In Stock
to see stock.
Add to basket
 
10259574
View Documents
Cytiva
28-9513-78
4 x 500 μL 8 to 17mg Human IgG/mL Native Protein A Protein A Mag Sepharose
154.00 €
Pack of 4
Estimated Shipment: 09-12-2024
to see stock.
Add to basket
 
Description

Description

Protein A and Protein G Mag Sepharose are magnetic beads designed to simplify enrichment of target proteins by immunoprecipitation or pull-down applications. The magnetic beads are based on Sepharose with native protein A or native protein G as ligand. Protein A and Protein G have high affinities for the Fc region of IgG antibodies and offer differing affinity characteristics across a broad range of mammalian IgG species.

There are two protocols available for protein enrichment: the cross-link protocol and the classic protocol. The protocols are simple with elution conditions optimized for for downstream applications such as electrophoresis and MS.

Protein A and Protein G Mag Sepharose combine well-proven enrichment methods with the magnetic beads platform, which has ideal properties for small-scale sample preparation. Together with MagRack 6, a separation tool for six microcentrifuge tubes, up to six samples can be processed in parallel.

  • Visible and dense Sepharose based magnetic beads with protein A or protein G ligand, easy to spot and collect bound target protein
  • Non-adherent beads eliminate smearing effects and aggregate formation. Can be used without detergents
  • Simple capture of target protein in small or large sample volumes (low microliter to high milliliter scale)
  • Optimized capacity for enrichment/immunoprecipitation of target proteins - only low amounts of antibody needed

Matrix: Paramagnetic, spherical, highly cross-linked agarose particles
Ligand: Native protein A; Native protein G
Binding capacity Protein A: 8-17mg human IgG/mL gel
Binding capacity Protein G: 13-22mg human IgG/mL gel
Particle size: 37-100μm
Working temperature: Room temperature
Storage solution: 20% ethanol
Storage temperature: 4°C to 8°C

Specifications

Specifications

Paramagnetic spherical, highly cross linked agarose particles
37 to 100 μm
4°C to 8°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Protein A Mag Sepharose, 1 x 500 ul

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3

Hazard Statement
  • H226-Flammable liquid and vapour.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/and all material-handling/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P235-Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanol
  • UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.

SDS
Documents

Documents

Product Certifications