missing translation for 'onlineSavingsMsg'
Learn More

Novex™ Protein A - Sepharose™ 4B

Product Code. 10727673
Click to view available options
Quantity:
5 mL
25 mL
1 L
Unit Size:
1 L
25mL
5mL
This item is not returnable. View return policy

Product Code. 10727673

Brand: Novex™ 101041

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protein A - Sepharose™ 4B

This product is prepared by covalently coupling highly purified (>98% by SDS-PAGE) Staph. aureus Cowan strain 1 Protein A to Sepharose™ 4B.

This product is supplied as a 50% suspension consisting of Protein A-Sepharose™ 4B and PBS (10 mM phosphate-buffered saline, pH 7.4, containing 0.1% Proclin).

Applications: Immunoglobulin Affinity Purification, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein A Sepharose
Product Line Sepharose
Column Type Affinity
Stationary Phase Protein A
Content And Storage This product is supplied as a 50% suspension (5 ml Protein A-Sepharose™ 4B + 5 ml PBS) Wet Ice
Quantity 5 mL
Form Suspension
compliance-icons
Product Identifier
  • Protein A - Sepharose 4B
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Proclin

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.